Learn More
Description
Specifications
Specifications
| Antigen | Influenza A H1N1 Nucleoprotein |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 24086762), Immunohistochemistry-Paraffin reported in scientific literature (PMID 16873262) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | NP_001081025.1 |
| Host Species | Rabbit |
| Immunogen | Recombinant Influenza A H1N1 Nucleoprotein. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
