Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody (2F205), HRP, Novus Biologicals™
SDP

Catalog No. NBP224871H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224871H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224871H Supplier Novus Biologicals Supplier No. NBP224871H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A nucleoprotein Monoclonal specifically detects Influenza A nucleoprotein in Virus samples. It is validated for ELISA.

Specifications

Antigen Influenza A nucleoprotein
Applications ELISA
Classification Monoclonal
Clone 2F205
Conjugate HRP
Dilution ELISA
Gene Accession No. O73619
Gene Alias Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Host Species Mouse
Immunogen Recombinant influenza A protein was used as the immunogen
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 394335
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.