Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody (2F205), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337292 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337292 Supplier Novus Biologicals Supplier No. NBP224871MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H1N1 Nucleoprotein Monoclonal antibody specifically detects Influenza A H1N1 Nucleoprotein in Virus samples. It is validated for ELISA

Specifications

Antigen Influenza A H1N1 Nucleoprotein
Applications ELISA
Classification Monoclonal
Clone 2F205
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant influenza A H1N1 protein was used as the immunogen
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.