Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody - (A/WSN/1933), Novus Biologicals™
SDP

Catalog No. NBP216965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216965 Supplier Novus Biologicals Supplier No. NBP216965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A nucleoprotein Polyclonal specifically detects Influenza A nucleoprotein in Viral samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA.

Specifications

Antigen Influenza A nucleoprotein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:20000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation -Assay dependent, Immunohistochemistry-Paraffin -Assay dependent, Sandwich ELISA -Assay dependent
Formulation PBS pH7, 20% Glycerol with 0.025% Proclin 300
Gene Accession No. H1N1
Gene Alias Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Host Species Rabbit
Immunogen Recombinant protein of Influenza A virus Nucleoprotein (A/WSN/1933(H1N1). The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.