Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody (F8), Novus Biologicals™
SDP

Catalog No. NB200414 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200414 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200414 Supplier Novus Biologicals Supplier No. NB200414

Mouse Monoclonal Antibody

Influenza A H1N1 Nucleoprotein Monoclonal antibody specifically detects Influenza A H1N1 Nucleoprotein in Influenza A Virus H1N1, Virus samples. It is validated for ELISA, Immunocytochemistry.

Specifications

Antigen Influenza A H1N1 Nucleoprotein
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone F8
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry 1:50-1:200
Gene Accession No. P03466
Gene Alias HA, Hemagglutinin
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified influenza virus type A strain H1N1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956529
Test Specificity This reacts with nucleoprotein (NP), which is common for Influenza A(H1N1), A(H3N2) and other Influenza A viruses. This antibody detects NP of Influenza A with high specificity and does not have cross reactivity to NP of Influenza B virus.
Target Species Influenza virus, Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.