Learn More
Influenza A H1N1 PA Antibody - (A/Puerto Rico/8/1934), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifically detects Influenza A Virus PA in Monkey, Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | Influenza A H1N1 PA |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300 |
| Gene Alias | Influenza PA |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus PA (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary. |
| Molecular Weight of Antigen | 83 kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.