Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H5N1 Hemagglutinin Antibody (1E6A7), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358714 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358714 Supplier Novus Biologicals Supplier No. NBP175493MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E6A7
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.