Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H5N1 Hemagglutinin Antibody (4E11E1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP175505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175505 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175505 Supplier Novus Biologicals Supplier No. NBP175505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Influenza A Hemagglutinin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4E11E1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.5 ug/ml, ELISA 2 ug/mL
Gene Accession No. AAT76166
Gene Alias HA epitope tag, HA1, HA2, Hemagglutinin
Gene Symbols HA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 17 - 338 of H5 HA1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Isotype IgG1, IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.