Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H5N1 Hemagglutinin Antibody (92)
SDP

Catalog No. NB020994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020994 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB020994 Supplier Novus Biologicals Supplier No. NBP289757

Rabbit Monoclonal Antibody

Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza A Virus H5N1 samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 92
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias HA, Hemagglutinin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H5N1 (Avian Flu) Hemagglutinin extracellular domain.
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3654620
Target Species Influenza A virus H5N1, Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.