Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H5N1 Hemagglutinin Rabbit anti-Influenza virus, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB186664 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186664 100 μg
NB186665 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB186664 Supplier Novus Biologicals Supplier No. NBP312799100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Formulation 0.2μm filtered solution in PBS without Preservative
Host Species Rabbit
Immunogen Purified, recombinant Influenza A H5N1 Hemagglutinin (AAT73273.1; Met1-Gln531)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Influenza virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.