Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H6N2 Hemagglutinin Antibody (13) - (A/chicken/Guangdong/C273/2011) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB306060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306060 Supplier Novus Biologicals Supplier No. NBP323536
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H6N2 Hemagglutinin Monoclonal antibody specifically detects Influenza A H6N2 Hemagglutinin in Virus samples. It is validated for Western Blot

Specifications

Antigen Influenza A H6N2 Hemagglutinin
Applications Western Blot
Classification Monoclonal
Clone 13
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation 0.2 um filtered solution in PBS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H6N2 Hemagglutinin (A/chicken/Guangdong/C273/2011) (Accession#: AFJ19048.1; Met1-Arg344)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.