Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H7 Hemagglutinin Antibody (InA414), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB358729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358729 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358729 Supplier Novus Biologicals Supplier No. NBP150817DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H7 Hemagglutinin Monoclonal antibody specifically detects Influenza A H7 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA, ELISA, Direct ELISA

Specifications

Antigen Influenza A H7 Hemagglutinin
Applications Western Blot, ELISA, Sandwich ELISA, Direct ELISA
Classification Monoclonal
Clone InA414
Conjugate DyLight 594
Dilution Western Blot, ELISA, Sandwich ELISA, Direct ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7).
Purification Method Protein A purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.