Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H7N1 Antibody (InA334), FITC, Novus Biologicals™
SDP

Catalog No. NBP150816F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150816F 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP150816F Supplier Novus Biologicals Supplier No. NBP150816F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H7N1 Monoclonal specifically detects Influenza A H7N1 in Virus samples. It is validated for ELISA.

Specifications

Antigen Influenza A H7N1
Applications ELISA
Classification Monoclonal
Clone InA334
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7).
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.