Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Haemagglutinin H5 Mouse anti-Virus, Biotin, Clone: 15A6, Novus Biologicals™
SDP

Catalog No. NB001894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001894 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001894 Supplier Novus Biologicals Supplier No. NBP105464B200UL

Mouse Monoclonal Antibody

Influenza A Haemagglutinin H5 Monoclonal specifically detects Influenza A Haemagglutinin H5 in Virus samples. It is validated for ELISA, Immunoassay, Sandwich ELISA.

Specifications

Antigen Influenza A Haemagglutinin H5
Applications ELISA
Classification Monoclonal
Clone 15A6
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias H5HA, Hemagglutinin 5
Host Species Mouse
Immunogen react with hemaglutinin H5 in hemagluttinin inhibition (HI) test
Quantity 200 μL
Test Specificity react with hemaglutinin H5 in hemagluttinin inhibition (HI) test
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.