Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A M1 Antibody (1G1A12), HRP, Novus Biologicals™
SDP

Catalog No. NBP250480H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250480H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250480H Supplier Novus Biologicals Supplier No. NBP250480H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A M1 Monoclonal antibody specifically detects Influenza A M1 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Influenza A M1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1G1A12
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Host Species Mouse
Immunogen GST-Influenza A M1 fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.