Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A M1/M2 Mouse anti-Virus, Clone: E10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF216525UL
Description
Influenza A M1/M2 Monoclonal antibody specifically detects Influenza A M1/M2 in Virus samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications
Influenza A M1/M2 | |
Monoclonal | |
0.5 mg/mL | |
Purified mouse monoclonal antibody IgG2a in PBS without azide. | |
KLH-conjugated linear peptide corresponding to 11 amino acids from M2 protein of Influenza A virus that protrudes out of the viral and cellular membrane. | |
25 μL | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
E10 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction