Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A M1/M2 Mouse anti-Virus, Clone: E10, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $468.00
Specifications
Antigen | Influenza A M1/M2 |
---|---|
Clone | E10 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF216525
|
MilliporeSigma
MABF216525UL |
25 μL |
Each for $182.31
|
|
|||||
MABF2165100
|
MilliporeSigma
MABF2165100UL |
100 μL |
Each for $468.00
|
|
|||||
Description
Influenza A M1/M2 Monoclonal antibody specifically detects Influenza A M1/M2 in Virus samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blotting.Specifications
Influenza A M1/M2 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2a in PBS without azide. | |
IgG2a κ | |
Protein G purified |
E10 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
KLH-conjugated linear peptide corresponding to 11 amino acids from M2 protein of Influenza A virus that protrudes out of the viral and cellular membrane. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title