Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A nucleoprotein Antibody (010), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB383176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383176 Supplier Novus Biologicals Supplier No. NBP312741UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Influenza A virus H3N2 samples. It is validated for Western Blot, ELISA

Specifications

Antigen Influenza A nucleoprotein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 010
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (Uniprot#: P22435.2; Met1-Asn498).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Influenza A virus H3N2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.