Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A nucleoprotein Antibody (1331), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N106046A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106046A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106046A94 Supplier Novus Biologicals Supplier No. NB10065046AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A nucleoprotein Monoclonal specifically detects Influenza A nucleoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen Influenza A nucleoprotein
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1331
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Flu
Host Species Mouse
Immunogen From Texas strain
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for the nucleoprotein of numerous strains and hence gives a ′nuclear′ staining in IF.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.