Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A nucleoprotein Antibody (7B4G10G8), Novus Biologicals™
SDP

Catalog No. NB287502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287502 20 μg
NB287399 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB287502 Supplier Novus Biologicals Supplier No. NBP32345220UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Influenza A nucleoprotein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7B4G10G8
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.