Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A nucleoprotein Antibody (7B4G10G8), Novus Biologicals™

Mouse Monoclonal Antibody
$170.70 - $463.50
Specifications
Antigen | Influenza A nucleoprotein |
---|---|
Clone | 7B4G10G8 |
Dilution | Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza A nucleoprotein | |
Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
7B4G10G8 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title