Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B Hemagglutinin Antibody (004)
SDP

Catalog No. NB021016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021016 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB021016 Supplier Novus Biologicals Supplier No. NBP289770

Rabbit Monoclonal Antibody

Influenza B Hemagglutinin Monoclonal specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Influenza B Hemagglutinin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias B1AG46
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Influenza B Hemagglutinin (ACA33493.1; Met 1-Ala 555) .
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Has cross-reactivity in ELISA and WB with: Influenza B (B/Brisbane/60/2008) HA Influenza B (B/Colorado/06/2017) HA Influenza B (B/PHUKET/3073/2013) HA Influenza B (B/Washington/02/2019) HA Has cross-reactivity in ELISA with: H1N1 (A/California/07/2009) HA Medium/Minimal cross-reactivity in ELISA with: H3N2 (A/Hong Kong/2671/2019) HA No cross-reactivity in ELISA and WB with: H1N1 (A/Brisbane/02/2018) HA H1N1 (A/California/04/2009) HA H1N1 (A/California/07/2009) HA H1N1 (A/Guangdong-Maonan/SWL1536/2019)/ (A/Hawaii/70/2019) HA H1N1 (A/Michigan/45/2015) HA H1N1 (A/Puerto Rico/8/1934) HA H1N1 (A/Wisconsin/588/2019)/ (A/Victoria/2570/2019) HA H3N2 (A/Brisbane/10/2007) HA H3N2 (A/Cambodia/e0826360/2020) HA H3N2 (A/Hong Kong/45/2019) HA H3N2 (A/Hong Kong/4801/2014) HA H3N2 (A/Kansas/14/2017) HA H3N2 (A/Singapore/INFIMH-16-0019/2016) HA H3N2 (A/Switzerland/9715293/2013) HA No cross-reactivity in ELISA with: H2N2 (A/Canada/720/2005) HA H4N6 (A/mallard/Ohio/657/2002) HA H5N1 (A/Anhui/1/2005) HA H6N1 (A/northern shoveler/California/HKWF115/2007) HA H7N7 (A/Netherlands/219/03) HA H8N4 (A/pintail duck/Alberta/114/1979) HA H9N2 (A/Hong Kong/1073/99) HA H10N3 (A/duck/Hong Kong/786/1979) HA H11N2 (A/duck/Yangzhou/906/2002) HA H12N5 (A/green-winged teal/ALB/199/1991) HA H13N8 (A/black-headed gull/Netherlands/1/00) HA H15N8 (A/duck/AUS/341/1983) HA H16N3 (A/black-headed gull/Sweden/5/99) HA No cross-reactivity in WB with: H3N2 (A/Hong Kong/2671/2019) HA
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.