Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B Hemagglutinin Antibody, DyLight 350 - (B/Florida/4/2006), Novus Biologicals™
SDP

Catalog No. NB182462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182462 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182462 Supplier Novus Biologicals Supplier No. NBP306576UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Influenza B Hemagglutinin Polyclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Influenza B Hemagglutinin
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias B1AG46
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Influenza B virus (B/Florida/4/2006) Hemagglutinin (ACA33493.1; Met1-Ala555).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.