Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B nucleoprotein Antibody (1D8) - (B/Lee/40) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP22351401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22351401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22351401 Supplier Novus Biologicals Supplier No. NBP2235140.1ML

Mouse Monoclonal Antibody

Influenza B Virus Nucleoprotein Monoclonal specifically detects Influenza B Virus Nucleoprotein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Influenza B Virus Nucleoprotein
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1D8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias Influenza B Virus NP, NP, Nucleoprotein
Host Species Mouse
Immunogen Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37C.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Clone 1D8 reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Clone 1D8 does not cross-react with influenza A virus.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.