Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza M1/ HLA-A2 Complex Antibody (405H1.01), Biotin, Novus Biologicals™
SDP

Catalog No. DDX0270B100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0270B100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0270B100 Supplier Novus Biologicals Supplier No. DDX0270B100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza M1 / HLA-A2 Complex Monoclonal specifically detects Influenza M1 / HLA-A2 Complex in Virus samples. It is validated for ELISA.

Specifications

Antigen Influenza M1 / HLA-A2 Complex
Applications ELISA
Classification Monoclonal
Clone 405H1.01
Concentration 0.5mg/mL
Conjugate Biotin
Dilution ELISA 1:100-1:2000
Gene Alias Influenza M1/HLA A2 Complex
Host Species Mouse
Immunogen Influenza M1/HLA-A2 Complex was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.