Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600908 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600908 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600908 Supplier Novus Biologicals Supplier No. NB600908
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ING1 Polyclonal specifically detects ING1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ING1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:4000-1:14000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9UK53
Gene Alias growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth family, member 1, inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, tumor suppressor ING1
Gene Symbols ING1
Host Species Goat
Immunogen This affinity purified ING1 Antibody was prepared from whole Goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 275-300 of Human ING1 protein (Inhibitor of growth family, member 1). (Uniprot: Q9UK53)
Molecular Weight of Antigen 46.74 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3621
Test Specificity Reactivity occurs against human p33 ING11 protein and is 100% homologous to all splice variants for ING11. Cross reactivity with p33 ING11 protein from mouse and rat sources is expected based on high homology (one amino acid change in sequence) by BLAST. Reactivity with p33 ING11 proteins from other sources is not known.
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.