Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING2 Antibody (AT39E5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB163232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163232 Supplier Novus Biologicals Supplier No. NBP261159IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ING2 Monoclonal antibody specifically detects ING2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen ING2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone AT39E5
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ING1Linhibitor of growth family, member 1-like, ING1Lp, Inhibitor of growth 1-like protein, inhibitor of growth family, member 2, inhibitor of growth protein 2, p33ING2p32
Host Species Mouse
Immunogen Recombinant human ING2 (1-280aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3622
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.