Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING3 Goat anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB029843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029843 Supplier Novus Biologicals Supplier No. NBP178100100UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ING3 Polyclonal specifically detects ING3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ING3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:2000, ELISA 1:10000 - 1:40000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Accession No. NP_061944
Gene Alias FLJ20089, ING2, inhibitor of growth family, member 3, inhibitor of growth protein 3, MEAF4, P47, p47ING3Eaf4
Host Species Goat
Immunogen This affinity purified antibody was prepared from whole Goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 294-304 of Human ING3 protein (Inhibitor of growth family, member 3). This sequence only shows homology to isof
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54556
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.