Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING4 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB151177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151177 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151177 Supplier Novus Biologicals Supplier No. NB1001682IR
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen ING4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein
Host Species Goat
Immunogen This affinity purified ING4 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4. (Uniprot: Q9UNL4)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51147
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.