Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin alpha Antibody (OTI12F11), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271039R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271039R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271039R Supplier Novus Biologicals Supplier No. NBP271039R

Mouse Monoclonal Antibody

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen Inhibin alpha
Applications Western Blot
Classification Monoclonal
Clone OTI12F11
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Gene Symbols INHA
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 62-232 of human Inhibin alpha (NP_002182) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3623.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.