Learn More
Description
Specifications
Specifications
| Antigen | Inhibin alpha |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | PO12/8 |
| Conjugate | DyLight 350 |
| Dilution | ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha |
| Gene Symbols | INHA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
