Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin alpha Antibody (PO12/8), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB308584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308584 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308584 Supplier Novus Biologicals Supplier No. NBP261936MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Inhibin alpha
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone PO12/8
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Host Species Mouse
Immunogen Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of Inhibin alpha
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3623
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.