Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (SR1672), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32185950UL
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Inhibin alpha | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1672 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthesized peptide derived from human Inhibin alpha (Uniprot #: P05111) | |
50 μg | |
Apoptosis, Cancer | |
3623 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction