Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (SR1672), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Inhibin alpha |
---|---|
Clone | SR1672 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Inhibin alpha | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthesized peptide derived from human Inhibin alpha (Uniprot #: P05111) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1672 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3623 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title