Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Mouse anti-Human, Clone: INHA/4266, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30745820UG
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Inhibin alpha | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthetic peptide (around aa73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary) | |
20 μg | |
Apoptosis, Cancer | |
3623 | |
Store at 4°C. | |
IgG2a |
Immunohistochemistry (Paraffin) | |
INHA/4266 | |
Unconjugated | |
1 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction