Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Mouse anti-Human, Clone: INHA/4266, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | Inhibin alpha |
|---|---|
| Clone | INHA/4266 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Inhibin alpha | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer | |
| 1 mM PBS with 0.05% BSA | |
| 3623 | |
| IgG2a | |
| Protein A or G purified |
| INHA/4266 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
| A synthetic peptide (around aa73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title