Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin beta A Rabbit anti-Human, Mouse, Rat, Clone: S03-1F0, Novus Biologicals™
SDP

Catalog No. NB188447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188447 25 μg
NB188446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188447 Supplier Novus Biologicals Supplier No. NBP31504725UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Inhibin beta A Monoclonal antibody specifically detects Inhibin beta A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Inhibin beta A
Applications Western Blot
Classification Monoclonal
Clone S03-1F0
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1
Host Species Rabbit
Immunogen A synthetic peptide of human Inhibin beta A (Uniprot#: P08476)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3624
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.