Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin beta B Antibody (34B10), Novus Biologicals™
SDP

Catalog No. NB298579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298579 100 μL
NB298578 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298579 Supplier Novus Biologicals Supplier No. NBP326199100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Inhibin beta B Monoclonal antibody specifically detects Inhibin beta B in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Inhibin beta B
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 34B10
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias activin AB beta polypeptide, Activin beta-B chain, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2, MGC157939
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Inhibin beta B [UniProt P09529]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3625
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.