Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Ink Chromatography and Forensics STEM Kit
SDPSCI_ED

Catalog No. s07360 Shop All United Scientific Supplies, Inc Products
Click to view available options
Class Size:
40 Students

Includes

  • Materials for three detailed activities (with model and inquiry investigational paths – a total of four investigational activities), which can accommodate up to 40 students
  • DVD with PDF teacher and student guides
  • Structured, guided and open investigations at introductory, intermediate and advanced levels

Requires

Additional household materials for independent investigations

The Ink Chromatography and Forensics STEM Kit introduces students to ink chromatography and its application in solving forensics cases.

Students gain experience in creating ink databases, identifying “ink tags” to “date” various inks and then use them (as professional document examiners do) in solving two mysteries involving document inks — an apparent suicide note and a forgery case.

  • Covers key concepts, including: data analysis, chromatographs, inks and their composition-mixtures, compounds, mixtures, molecules, solubility, affinity, polar substances, Rf values, capillary action, light energy, fluorescence and hypothesis testing
  • Accents the scientific method and independent inquiry featuring hands-on learning materials and exciting interactive digital content that can be accessed on a computer, tablet or mobile device
  • Provides access to digital PDF content, such as: independent investigation inquiry suggestions; background information including content tables and concept; animations; PowerPoint™ concept presentation (and video version); comprehensive glossary
  • Special typewriter functionality in student activity PDFs allows students to take notes, record experimental data, answer analysis questions and e-mail their teacher
  • Correlated to the Next Generation Science Standards

Specifications

Grade 5 to 12
For Use With (Application) Introduces Students to Ink Chromatography and Its Application in Solving Forensics Cases
Includes Materials for three detailed activities (with model and inquiry investigational paths - a total of four investigational activities), which can accommodate up to 40 students, DVD with PDF teacher and student guides, Structured, guided and open investigations, (at introductory, intermediate and advanced levels)
Class Size 40 Students
Product Type Ink Chromatography and Forensics Stem Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.