Learn More
Innocal™ Humidity Calibrations
Keeping humidity instruments calibrated with InnoCal™ Humidity Calibrations helps measure and regulate this variable more accurately.
$444.93 - $582.00
Specifications
Certifications/Compliance NIST-Traceable
Duration 12 Months
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
15-094-31
View Documents
Innocal™
WD1700500
Humidity (Psychrometer)
Each for $444.93
Only null left
Add to Cart
15-094-37
View Documents
Innocal™
WD1703020
Humidity (High Accuracy)
Each for $582.00
Only null left
Add to Cart
Description
Description
  • All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu's), and test uncertainty ratios (TUR's)
  • NIST-traceable calibration allows users to confirm that the specific asset purchased meets the manufacturer's published specification
  • EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
  • Product arrives calibrated and ready to use avoiding the necessity of sending it to a third-party or internal calibration lab
  • All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A.
Specifications
Specifications
NIST-Traceable
12 Months
Videos
SDS
Documents
Documents
Product Certifications
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.