Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ InnovaCoat (R) GOLD 10nm Mini Antibody Labeling Kit
Shop All R&D Systems Products

Click to view available options
Quantity:
10 Tests
3 Tests
Description
Features
- Proprietary surface coat reduces risk of sub-optimal antibody performance
- Ultra-stable
- Covalently link antibodies and analytes
- Simple conjugation processes
- Different surface chemistries
- Antibody-GOLD conjugates in just 15 minutes
- Enhanced assay sensitivity
- Protective coat prevents metal-antibody interactions
- Coating procedure not dependent on isoelectric point of antibody
- Rapid screening of multiple antibodies for assay development
- Reduced antibody loading without aggregation
- No pH titrations
- Fully scalable – from R&D to production
- Custom InnovaCoat™ labeling services
Specifications
Specifications
| Quantity | 10 Tests |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Type | Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction