Learn More
Novus Biologicals™ InnovaCoat (R) GOLD 40nM Midi Antibody Labeling Kit
Shop All R&D Systems Products
Click to view available options
Quantity:
1 Test
Description
Features
- Proprietary surface coat reduces risk of sub-optimal antibody performance
- Ultra-stable
- Covalently link antibodies and analytes
- Simple conjugation processes
- Different surface chemistries
- Antibody-GOLD conjugates in just 15 minutes
- Enhanced assay sensitivity
- Protective coat prevents metal-antibody interactions
- Coating procedure not dependent on isoelectric point of antibody
- Rapid screening of multiple antibodies for assay development
- Reduced antibody loading without aggregation
- No pH titrations
- Fully scalable – from R&D to production
- Custom InnovaCoat™ labeling services
Specifications
Specifications
| Content And Storage | -20°C |
| Quantity | 1 Test |
| Product Type | Antibody Labeling Kit |