Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD - 40nm Mini Maleimide Antibody Labeling Kit
SDP

Catalog No. 2700005 Shop All R&D Systems Products
Click to view available options
Quantity:
7.5 μg
For Research Use Only

Catalog No. 27-000-05

Supplier: Novus Biologicals™ 2700005

Only null left
Add to Cart
Add to Cart
For Research Use Only

Use simple pipetting to covalently attach thiolated molecules to ultra-stable InnovaCoat™ GOLD nanoparticles. Simply add solution of thiolated biomolecule to the supplied freeze-dried mixture. Attached molecule isn'tt prone to dissociation, unlike many direct thiol-to-gold metal conjugations.

Features
  • Proprietary surface coat
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services

Specifications

Quantity 7.5 μg
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.