Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) Gold - 20nm Midi Maleimide Antibody Labeling Kit
SDP

Catalog No. p-7112403 Shop All R&D Systems Products
Click to view available options
Sufficient For:
1 Reaction
3 Reactions

20nm malemide nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • Allows thiolated molecules to be covalently attached to nanoparticles
  • Shields biomolecule from metal surface thus eliminating metal-antibody interactions
  • Reduces risk of sub-optimal antibody performance

Applications: Electron microscopy visualization
Storage: Store at 4°C

Note: Kits are guaranteed for 6 months from date of receipt

Specifications

Quantity 50 μg
Product Type 20nm Mini Maleimide Antibody Labeling Kit
Sufficient For 1 Reaction
Content And Storage -20°C

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.