Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers INNOVANCE™ VWF Ac (von Willebrand Activity) Assay Kit

Catalog No. 23044883 Shop All Siemens Healthineers Products
Change view
Click to view available options
No. of Reactions:
∼110 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
23-044-883 ∼110 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Materials required but not provided with the kit: Standard Human Plasma, Control Plasma N, Control Plasma P and Owren’s Veronal Buffer
Catalog No. 23-044-883 Supplier Siemens Healthineers Supplier No. 10487040
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Materials required but not provided with the kit: Standard Human Plasma, Control Plasma N, Control Plasma P and Owren’s Veronal Buffer

ISTH guideline recommended functional assay for detecting quantitative and qualitative VWF defects, using no Ristocetin

  • Insensitive to VWF polymorphisms, which could lead Ristocetin assays to erroneous diagnosis of VWD
  • Liquid, ready to use reagent
  • Measuring range of 4 to 300%, with <5% precision at Medical Decision Point
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Human Plasma
Type Coagulation Assay
Includes Reagent 1 (3 vials), Reagent 2 (3 vials) and Reagent 3 (1 vial)
No. of Reactions ∼110 Tests
Range 4 to 300% of norm
Sample Size 16 μL
Stability 24 to 72 hours (Sysmex CS-2500 and CS-5100 Systems); 36 hours (BCS™ XP System)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.