Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INO80 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290838 Supplier Novus Biologicals Supplier No. NBP178758F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INO80 Polyclonal antibody specifically detects INO80 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen INO80
Applications Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias EC 3.6.1, EC 3.6.1.23, EC 3.6.4.12, hINO80INO80 complex subunit A, homolog of yeast INO80, Ino80, INO80 complex homolog 1 (S. cerevisiae), INO80 homolog (S. cerevisiae), INO80A, INOC1putative DNA helicase INO80 complex homolog 1, KIAA1259
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1400 and 1450 of human INO80 homolog using the numbering given in entry NP_060023.1 (GeneID 54617).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54617
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.