Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INO80E Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB054413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054413 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054413 Supplier Novus Biologicals Supplier No. NBP298592FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INO80E Polyclonal specifically detects INO80E in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen INO80E
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CCDC95, coiled-coil domain containing 95, Coiled-coil domain-containing protein 95, FLJ00079, FLJ90652, INO80 complex subunit E
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human INO80E fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283899
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.