Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inorganic Pyrophosphatase/PPA1 Antibody (4G4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB162027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162027 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162027 Supplier Novus Biologicals Supplier No. NBP259471IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Inorganic Pyrophosphatase/PPA1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 4G4
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061
Host Species Mouse
Immunogen Recombinant human Inorganic Pyrophosphatase/PPA1 (1-289aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5464
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.