Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iNOS Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB327119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327119 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327119 Supplier Novus Biologicals Supplier No. NBP299091H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

iNOS Polyclonal antibody specifically detects iNOS in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen iNOS
Applications Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human iNOS.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4843
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.