Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Inosine, 98+%

Catalog No. p-7021925
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol

Suppresses the increase of glucose and insulin in the blood

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
MDL Number MFCD00066770
InChI Key UGQMRVRMYYASKQ-YPLCUDRINA-N
Synonym inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
PubChem CID 6021
ChEBI CHEBI:17596
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 212°C to 215°C (decomposition)
Quantity 5 g
Beilstein 624889
Merck Index 14,4975
Optical Rotation −52° (c=1 in Water)
Formula Weight 268.23
Percent Purity ≥98%
Chemical Name or Material Inosine
EINECSNumber 200-390-4
RTECSNumber NW7460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.